Learn More
BTX™ Electroporation Accessories: Banana Plug Connector
Use the Banana Plug Connector to connect external devices (Genetrodes, Genepaddles, Microslides, Meander Fusion Chamber) to BTX electroporation generators.
Brand:  BTX™ 450216
Product Code. 15457300
  • £285.00 / Each
Explore more special offers
Description
Description
  • Compatible with most BTX Systems such as ECM 2001+ and Gemini
  • See the electrode type for more details
Specifications
Specifications
Connect The Genetrode, Genepaddle, Microslide, Meander Fusion Chamber to The Electroporation Generator
BTX Electroporation
10 ft.
Non-sterile
Banana Plug to Micrograbber
ECM 630, ECM 830, Gemini X2, Legacy ECM 2001 Electroporation Generators
Micrograbber to Banana Plug Connection Cables, 10 ft
3.1 m
Electroporation Accessory
AC Sine Wave, DC Square Wave