missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Legacy ECM 830 Foot Switch (compatible with non-touchscreen models)

Product Code. 15407300
Click to view available options
Connection Type:
Output Cable
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15407300

Brand: BTX™ 450211

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Legacy ECM 830 Foot Switch allows hands-free operation.

  • Functions in place of the start button on the front of the generator
  • Ideal when conducting in vivo/in ovo gene delivery or nuclear transfer experiments when both hands are needed for sample manipulation
  • Compatible with BTX generators as specified in table1

1Also compatible with some older model units. Contact your Fisher Scientific™ Customer Service Representative for more information.

Specifications

For Use With (Application) In-Vivo Gene Delivery, in-Ovo Gene Delivery and Nuclear Transfer/Cloning
For Use With (Equipment) Legacy ECM 830 Square Wave Electroporation System (Non-touch screen models)
Product Line BTX Electroporation
Product Type Electroporation accessory
Description Legacy ECM 830 Foot Pedal (compatible with non-touchscreen models)
Depth (English) 3.5 in.
Depth (Metric) 89 mm
Height (English) 2 in.
Height (Metric) 51 mm
Length (English) Cable 10 ft.
Length (Metric) Cable 3.1 m
Width (English) 4.5 in.
Width (Metric) 114 mm
Sterility Non-sterile
Connection Type Output Cable
Interface Footswitch
Pulse Form Square Wave
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.