Learn More
BTX™ HT 25-well Electroporation Plates
HT 25-well Electroporation Plates for use with ECM 630 and ECM 830 Electroporation Systems.
Brand:  BTX™ 450466
 View more versions of this product
Product Code. 15427320
  • £94.45 / Each
Explore more special offers
Description
Description
  • 25-well disposable electroporation plate
  • Standard multi-well formats
  • Bio-compatible materials
Specifications
Specifications
25-well
ECM 630HT, ECM 830HT, Gemini X2 HT Electroporation System
BTX Electroporation
25-Well Disposable Electroporation Plate, 2 mm gap, 100 μl, coated, 1 plate
1 x plate with seal
0.078 in.
100 μL (Working)
Exponential Decay Wave, Square Wave
siRNA Transfection, Library Studies and Stem Cell Project
Medical grade polycarbonate, polished aluminium
Electroporation consumables
25-well Electroporation Plate
1 Ea.
2 mm
Sterile
Yes