missing translation for 'onlineSavingsMsg'
Learn More

BTX™ HT 25-well Electroporation Plates

Product Code. p-4901666
Click to view available options
Size (Metric) Gap:
2 mm
4 mm
Volume (Metric):
100 μL (Working)
200 μL
Quantity:
1 Ea.
6 Pk.
Unit Size:
Each
Pack of 6
This item is not returnable. View return policy

Product Code. 15497310

Brand: BTX™ 450462

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

HT 25-well Electroporation Plates for use with ECM 630 and ECM 830 Electroporation Systems.

  • 25-well disposable electroporation plate
  • Standard multi-well formats
  • Bio-compatible materials

Specifications

Format 25-well
For Use With (Application) Transfection/transformation of bacteria, plant and mammalian cells, siRNA Transfection, Library Studies and Stem Cell Project
For Use With (Equipment) ECM 830 HT Electroporation System, the ECM 630 HT Electroporation System, or the Gemini X2 HT Electroporation System
Material Medical grade polycarbonate, polished aluminum
Product Line BTX Electroporation
Product Type Electroporation consumables
Description 25-Well Disposable Electroporation Plate, 4 mm gap, 200 μl, 1 plate
Electroporation Chamber 25-well Electroporation Plate
Quantity 1 Ea.
Size (English) Gap 0.15 in.
Size (Metric) Gap 4 mm
Volume (Metric) 200 μL
Sterility Sterile
Pulse Form Exponential Decay Wave, Square Wave
High-throughput Compatibility Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.