missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Tweezertrode™ Electrode

Tweezertrode™ Electrode is a reusable, tweezer-style, in vivo electrode suitable for drug or gene delivery in animals.

Brand:  BTX™ 450489

 View more versions of this product

Product Code. 15417330

  • £1805.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel
  • Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
  • Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
  • Can be easily cleaned

Order Info

Tweerzertrode kits (Cat. No. BTX450486) and additional cable (BTX524) also available separately.

Specifications

Specifications

in vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery
Platinum
Tweezer Style Electrode
Platinum Tweezertrode, 5 mm Diameter
5 mm
0.47 in.
41°F to 104°F
1 μsec. to 200 msec.
1 Ea.
Adjustable. <1 mm to >2 cm
11.5 cm
Banana Cables
BTX 830, Gemini X2, ECM2001+, Legacy ECM 2001 Generator
BTX Electroporation
200 V
0.196 in.
Circular
11.9 mm
1 sec. to 99 msec.
5°C to 40°C
20 to 80%
Tweezer Style
Non-sterile
Square Wave
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Intended for Research use only. Not for use in diagnostic or clinical procedures.