missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Tweezertrode™ Electrode

Tweezertrode™ Electrode is a reusable, tweezer-style, in vivo electrode suitable for drug or gene delivery in animals.

Brand:  BTX™ 450119

 View more versions of this product

Product Code. 15487260

  • £1245.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel
  • Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
  • Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
  • Can be easily cleaned

Order Info

Tweerzertrode kits (Cat. No. BTX450486) and additional cable (BTX524) also available separately.

Specifications

Specifications

In vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery.
Stainless Steel
Tweezer Style Electrode
Stainless Tweezertrode Electrode, 10 mm Diameter
10 mm
10 μsec. to 99 msec. in PBS
Tweezer Style
Non-sterile
Square Wave
ECM 830, ECM 2001+, Legacy ECM2001, Gemini X2
BTX Electroporation
1000 V
0.393 in.
Circular
1 Ea.
11.5 cm
Banana to Pin Tip
Adaptor Cable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Intended for Research use only. Not for use in diagnostic or clinical procedures.