missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Tweezertrodes™ Electrodes Accessory: Cable
Cable for use with Tweezertrodes Electrodes.
Brand: BTX™ 450204
Code : NEW
Additional Details : Weight : 0.25000kg
Description
- For connection to electroporation generators
- 1.1 m in length
Specifications
In-Vivo Drug or Gene Delivery, Gene Therapy, Protein Delivery | |
Adapter Banana Plug Cables, Red and Black (for use with Tweezertrodes and Adherent Cell Electrode) | |
3.5 ft. | |
Non-sterile | |
Exponential Decay Wave, Square Wave |
BTX Tweezertrodes, L-Shaped Genetrode Electrodes, Adherent Cell Electrode | |
Red and Black Cable Set | |
1.1 m | |
Banana Cables |