missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Genetrodes™ Electrodes
Click to view available options
Electrode Shape:
Bent L-shaped
Straight
Length (Metric) Electrode:
1 mm
3 mm
5 mm
10 mm
Unit Size:
Each
Description
- Needle-style reusable electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
- Five models available
- Models 508 and 510 are straight with gold tips designed for general in vivo applications
- Models 512, 514, and 516 are L-shaped electrodes with gold tips designed for in vivo and in ovo electroporation
Order Info
Genetrode holder with shaft available separately (Cat. No. BTX515).
Specifications
Specifications
| For Use With (Application) | In-Vivo Gene Delivery, ex-Vivo Gene Delivery, in-Ovo Gene Delivery |
| For Use With (Equipment) | ECM 830, ECM 2001+, ECM 2001, and Gemini X2 |
| Material | Gold |
| Product Line | BTX Electroporators |
| Product Type | Electroporation accessory |
| Voltage | 200 V |
| Description | Genetrodes, 5 mm L-Shape (GOLD TIP) |
| Electrode Shape | Bent L-shaped |
| Includes | Genetrode only |
| Length (English) Electrode | 0.19 in. |
| Show More |
Intended for Research use only. Not for use in diagnostic or clinical procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction