missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Genetrodes™ Electrodes
Needle-style reusable Genetrodes™ Electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems.
Brand: BTX™ 450116
This item is not returnable.
View return policy
Description
- Needle-style reusable electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
- Five models available
- Models 508 and 510 are straight with gold tips designed for general in vivo applications
- Models 512, 514, and 516 are L-shaped electrodes with gold tips designed for in vivo and in ovo electroporation
Order Info
Genetrode holder with shaft available separately (Cat. No. BTX515).
Specifications
In-Vivo Gene Delivery, ex-Vivo Gene Delivery, in-Ovo Gene Delivery | |
Gold | |
Electroporation accessory | |
Genetrodes, 3 mm L-Shape (GOLD TIP) | |
Genetrode only | |
3 mm | |
1 sec. to 99 msec. | |
1 Ea. | |
1 to 10 mm | |
Non-sterile | |
Square Wave |
ECM 830, ECM 2001+, ECM 2001, and Gemini X2 | |
BTX Electroporators | |
200 V | |
Bent L-shaped | |
0.11 in. | |
∼1500+ Pulses | |
10 μsec. to 99 msec. in PBS | |
0.03 to 0.39 in. | |
Gold Tip | |
Banana Plug to Micrograbber |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BTX™ Genetrodes™ Electrodes > Electrodes; Tip: L-Shape; Length: 3 mm (0.11 in.)
Intended for Research use only. Not for use in diagnostic or clinical procedures.
Spot an opportunity for improvement?Share a Content Correction