Learn More
BTX™ Safety Stand for Electroporation
Safety Stand compatible with BTX™ Gemini X2, ECM 399, ECM 630 and ECM 830 Electroporation Systems.
Brand: BTX™ 450207
This item is not returnable.
View return policy
Description
- For use with disposable Fisher cuvettes
- Unique design ensures no current unless the lid is closed
- Up to two cuvettes can be electroporated simultaneously
- Separate designs for Gemini X2 and Gemini SC
Specifications
Disposable Cuvettes, Holds Various Types of Chambers/Two Cuvettes at The Same Time | |
BTX Electroporation | |
Safety Stand, Adjustable Gap (for use with Cuvettes) | |
1 Ea. | |
Banana Cables |
Gemini X2, ECM 399, ECM 630, ECM 830 | |
Safety Stand | |
4 mm Banana Cables | |
Non-sterile | |
Exponential Decay Wave, Square Wave |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
BTX™ Safety Stand for Electroporation > Safety Stand; For use with: Gemini X2, ECM 399, ECM 630, ECM 830
Spot an opportunity for improvement?Share a Content Correction