missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Two-Needle Array™ Electrode Kit
Two-Needle Array™ Electrode Kit designed for in vivo drug or gene delivery.
Brand: BTX™ 450168
Includes:
- Handle
- Six two-needle arrays
Description
- Consists of reusable array handle and sterile, disposable two-needle array
- Enables simple, inexpensive, safe direct injection of muscle tissue for plasmid DNA delivery
- Compatible with ECM™ 830, ECM 2001, ECM 630
- Supplied in convenient kit with Delrin handle and 20 mm stainless-steel needle array (pack of 6), or order components separately (match handle size to needle array size)
Two Sizes
- 5 mm array and handle recommended for small muscle masses such as mouse tibialis
- 10 mm size suitable for larger muscle masses such as rat gastronomes
Specifications
In-Vivo Drug or Gene Delivery, Muscle Gene Therapy | |
Stainless Steel, Delrin | |
Electroporation accessory | |
2-Needle Array Kit, 5 mm, Pkg. of 6, with Handle | |
6 x 5 mm 2-needle Arrays, 1 x Reusable Handle | |
20 mm | |
5°C to 40°C | |
5 mm | |
Banana Cables |
ECM 830, Gemini X2, ECM 2001, ECM 2001+Generator | |
BTX Electroporation | |
500 V | |
Straight/Needle | |
0.78 in. | |
10 μsec. to 99 msec. in PBS | |
0.19 in. | |
Non-sterile | |
Square Wave |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Intended for Research use only. Not for use in diagnostic or clinical procedures.
Spot an opportunity for improvement?Share a Content Correction