missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Two-Needle Array™ Electrodes
Two-Needle Array™ Electrodes designed for in vivo drug or gene delivery.
Brand: BTX™ 450120
This item is not returnable.
View return policy
Description
- Sterile, disposable two-needle array
- Enables simple, inexpensive, safe direct injection of muscle tissue for plasmid DNA delivery
- Compatible with ECM™ 830, ECM 2001, ECM 630
Two Sizes
- 5 mm array and handle recommended for small muscle masses such as mouse tibialis
- 10 mm size suitable for larger muscle masses such as rat gastronomes
Specifications
In-Vivo Drug or Gene Delivery, Muscle Gene Therapy | |
Medical grade Stainless Steel | |
Electroporation accessory | |
2-Needle Array, 10 mm Gap, Pkg. of 6 | |
Pk. 6 Needle array. Requires handle and connector cables | |
20 mm | |
6 Pk. | |
10 mm | |
Sterile | |
Square Wave |
ECM 830, ECM 2001+, ECM 630, Legacy ECM 2001 Generator | |
BTX Electroporators | |
500 V | |
Straight/Needle | |
0.78 in. | |
10 μsec. to 99 msec. in PBS | |
0.39 in. | |
Needle array | |
Banana Plug to Banana Plug | |
Handle, Connector Cables |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Intended for Research use only. Not for use in diagnostic or clinical procedures.
Spot an opportunity for improvement?Share a Content Correction