Learn More
BTZ043, MedChemExpress
MedChemExpress BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), with MICs of of 2.3 nM and 9.2 nM for M. tuberculosis H37Rv and Mycobacterium smegmatis, respectively.
Brand:  Medchem Express HY-13579
Encompass_Preferred
 View more versions of this product
Product Code. 16498437
  • £95.59 / 5mg
Explore more special offers
Description
Description
BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), with MICs of of 2.3 nM and 9.2 nM for M. tuberculosis H37Rv and Mycobacterium smegmatis, respectively.
Specifications
Specifications
BTZ043
Yellow
C17H16F3N3O5S
DMSO : 13.3 mg/mL (30.83 mM; Need ultrasonic and warming)
431.39
99.75%
Solid
1161233-85-7
Research
5 mg
O=C1N=C(N(CC2)CCC32OC[C@H](C)O3)SC4=C([N+]([O-])=O)C=C(C(F)(F)F)C=C14
431.39
Research
Cancer-programmed cell death
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
BTZ043, MedChemExpress