missing translation for 'onlineSavingsMsg'
Learn More

BU 224 hydrochloride, Tocris Bioscience™

Product Code. 18094832
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective I2 ligand. Putative antagonist

High affinity ligand for the imidazoline I2 binding site (Ki = 2.1 nM). Putative I2 antagonist; antagonizes the effects of imidazoline ligands on morphine antinociception. Produces ipsiversive rotational behavior in rats with a full 6-OHDA lesion of the nigrostriatal tract.

Chemical Identifiers

CAS 205437-64-5
Molecular Formula C12H12ClN3
Molecular Weight (g/mol) 233.699
InChI Key DDFHQXAQWZWRSQ-UHFFFAOYSA-N
PubChem CID 11957470
IUPAC Name 2-(4,5-dihydro-1H-imidazol-2-yl)quinoline;hydrochloride
SMILES C1CN=C(N1)C2=NC3=CC=CC=C3C=C2.Cl

Specifications

Quantity 10 mg
Synonym bu224 hydrochloride, bu 224 hydrochloride, 2-4,5-dihydroimidazol-2-yl quinoline hydrochloride, 2-4,5-dihydro-1h-imidazol-2-yl quinoline hydrochloride, bu224 hydrochloride, solid, 2-4,5-dihydroimidazol-2-yl-quinoline hydrochloride, 2-4,5-dihydro-1h-imidazol-2-yl quinoline hydrochloride 1:1
Formula Weight 233.7
Percent Purity >98%
Chemical Name or Material BU 224 hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'