missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BU 226 hydrochloride, Tocris Bioscience™
Potent, highly selective I2 ligand
Chemical Identifiers
CAS | 1186195-56-1 |
---|---|
Molecular Formula | C12H12ClN3 |
Molecular Weight (g/mol) | 233.699 |
InChI Key | NIGHNKWLMSRCOZ-UHFFFAOYSA-N |
Synonym | bu 226 hydrochloride, 2-4,5-dihydroimidazol-2-yl isoquinoline hydrochloride |
PubChem CID | 56972194 |
IUPAC Name | 3-(4,5-dihydro-1H-imidazol-2-yl)isoquinoline;hydrochloride |
SMILES | C1CN=C(N1)C2=CC3=CC=CC=C3C=N2.Cl |
Description
A high affinity ligand for I2 imidazoline sites (Ki = 2.7 nM) with very high selectivity (> 2000-fold) over α2-adrenoceptors (Ki = 6700 nM).Chemical Identifiers
1186195-56-1 | |
233.699 | |
bu 226 hydrochloride, 2-4,5-dihydroimidazol-2-yl isoquinoline hydrochloride | |
3-(4,5-dihydro-1H-imidazol-2-yl)isoquinoline;hydrochloride |
C12H12ClN3 | |
NIGHNKWLMSRCOZ-UHFFFAOYSA-N | |
56972194 | |
C1CN=C(N1)C2=CC3=CC=CC=C3C=N2.Cl |
Specifications
1186195-56-1 | |
bu 226 hydrochloride, 2-4,5-dihydroimidazol-2-yl isoquinoline hydrochloride | |
C1CN=C(N1)C2=CC3=CC=CC=C3C=N2.Cl | |
233.699 | |
233.7 | |
BU 226 hydrochloride |
C12H12ClN3 | |
NIGHNKWLMSRCOZ-UHFFFAOYSA-N | |
3-(4,5-dihydro-1H-imidazol-2-yl)isoquinoline;hydrochloride | |
56972194 | |
>98% |
Safety and Handling
Recommended Storage : Store at Room Temperature