Learn More
Notrax Bubble Sof-Tred™ Dyna-Shield™ Layer with Unique Bubble Design
Notrax Bubble Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.
Brand: Notrax 417C0048GY
Additional Details : Weight : 4.70000kg
Description
- Lengthwise corrugated rib pattern for sure footing
- Exceptional worker comfort due to 127 mm thick specially formulated compound
- Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl
- Available with yellow borders along sides comply with OSHA code 1910-144
- All 4 sides are bevelled and sealed to minimize tripping hazards
Specifications
60 ft. | |
Industrial and Commercial Matting | |
122 cm |
Gray | |
Microcellular Vinyl | |
1830 cm |