Learn More
Kinesis™ Bubble trap hi-flow vacuum PTFE and PEEK pressure rated to 30psi (2 bar) 1/4 inch-28 UNF threaded ports Omnifit™
Brand:  Kinesis™ 006BT-HF
Encompass_Preferred
Product Code. 13154044