missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Boiling Point Tubes and Capillaries

Product Code. p-4463121 Shop All Buchi Products
Click to view available options
Product Type:
Boiling Point Capillaries
Boiling Point Tubes
Packaging Quantity:
100/Pk.
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12030592

Brand: BUCHI 019007

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Boiling point tubes and capillaries for automated or manual determination according to the Siwoloboff method.

  • High precision for reliable results
  • Meets Siwoloboff and Eu Pharmacopeia standards
  • Designed for BUCHI melting and boiling point instruments
  • Compatible with other instruments

Specifications

Product Type Boiling Point Tubes
Diameter (Metric) 2.75 mm
For Use With (Application) For reproducible boiling point determinations
For Use With (Equipment) Boiling point apparatus
Material Borosilicate Glass 3.3
Packaging Quantity 100/Pk.
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.