Learn More
BUCHI Accessories for Pumps: Membrane Head
For use with V-700/V-710 vacuum pumps.
Brand: BUCHI 047153
Code : 61
Additional Details : Weight : 0.31020kg
Description
- PTFE-rubber composite membrane
- With structured surface for optimal vacuum generation
Specifications
Model V700/V710 vacuum pump | |
PTFE Rubber Composite Membrane | |
Membrane Head |