Learn More
BUCHI Rotavapor Bump Trap SJ 24/40
Bump trap adapter.
Brand: BUCHI 11056919
Additional Details : Weight : 0.22000kg
Description
- BUCHI glassware
- Glass adapters for excessively foaming samples
- Prevents foam from entering the vapor duct and condenser
- Glass, SJ24/40, 175mm
Specifications
Bump Trap | |
Modifies vapor pathway in event of bumping or foaming | |
Rotary evaporator | |
24/40 |