Learn More
BUCHI Agate Mortar and Pestle Set
Agate mortar and pestle for high purity sample crushing. The polished surface is easy to clean and prevents cross-contaminations.
Brand: BUCHI 041867
Additional Details : Weight : 0.07000kg
Description
- Sized for melting point sample preparation
- Ensure sample consistency and repeatable results
- The polished surface is easy to clean and prevents cross-contamination
Specifications
Polished | |
Mortar and Pestle | |
Agate Stone |