Learn More
BUCHI Condensers for BUCHI Rotavapors
Original BUCHI glassware guarantees the highest levels of efficiency and safety thanks to high precision in manufacturing. For more than 80 years, BUCHI has been manufacturing innovative and durable glass parts to ease daily laboratory work.
Brand: BUCHI 040651
Code : A0
Additional Details : Weight : 1.50000kg
Description
- High throughput due to advanced design of condensers
- Highly resistant glass provides the highest levels of safety (DURAN™ borosilicate glass 3.3)
- Guaranteed leak-tightness and protection against hazardous fumes thanks to high precision joints
- Maximum stability and safety with plastic and glass coating
Specifications
Condenser S | |
Evaporator Condensation | |
Rotavapor R-210/R-215 | |
None |