Learn More
BUCHI Glass Columns C-690
£1091.53 - £6710.00
Specifications
For Use With Preparative Separations Together with The C-600 Sepacore™ Line of Chromatography Separators
Material Borosilicate Glass
View More Specs
Products 8
Product Code Brand Length (Metric) Diameter (Metric) Max. Pressure Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Max. Pressure Price Quantity & Availability  
12765995
View accessory products View Documents
BUCHI
044036
230 mm 26 mm 580 psi
£1290.00
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
11967208
View Documents
BUCHI
044037
460 mm 26 mm 580 psi This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12739485
View accessory products View Documents
BUCHI
044045
460 mm 49 mm 290 psi
£1780.00
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12749485
View accessory products View Documents
BUCHI
044046
920 mm 49 mm 290 psi
£1965.00
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12769485
View accessory products View Documents
BUCHI
044049
460 mm 70 mm 218 psi
£2725.00
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12779485
View accessory products View Documents
BUCHI
044050
920 mm 70 mm 218 psi
£3350.00
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12799485
View accessory products View Documents
BUCHI
044053
460 mm 100 mm 145 psi
£5140.00
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12709495
View accessory products View Documents
BUCHI
044054
920 mm 100 mm 145 psi
£6710.00
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
Specifications
Preparative Separations Together with The C-600 Sepacore™ Line of Chromatography Separators
Borosilicate Glass