Learn More
BUCHI Ball Joint Clamp for BUCHI condenser and receiving flask
Clamp is designed to work with ground glass ball joints on BUCHI condensers and receiving flasks.
Brand: BUCHI 003275
Code : A0
Additional Details : Weight : 0.07000kg
Description
- Joint: SJ20/35
- Fits all models of condensers
Specifications
Stainless Steel | |
Joint Clamp | |
BUCHI Syncore™ Polyvap parallel evaporation systems | |
SJ35/20 |