missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Set of Verification Substances for Melting Point Apparatus

For use with Melting Point apparatus.

Brand:  BUCHI 037667


Product Code. 10453322

  • £704.00 / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket

Includes: 4-Acetylbenzonitrile, m-Toluic acid, Sulfanilamide and Phenolphthalein
This item is not returnable. View return policy

Description

Description

For use with Melting Point apparatus.
Specifications

Specifications

Verification Substances Set
B-545 melting point apparatus
4-acetylbenzonitrile, m-toluic acid, sulfanilamide and phenolphthalein
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • buchi B540 melting point calibration kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : phenacetin, caffeine, vanillin

SDS
Documents

Documents

Certificates
Special Offers

Special Offers