missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Rotation Drying Flask

Accessory for Kugelrohr™ Glass Oven B-585

Brand:  BUCHI 037143

Product Code. 10309023

  • £398.00 / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Joints are conically tapered, size 24/29
  • Flask for drying up to 30mL of solid substances

Specifications

Specifications

30 mL
Drying up to 30mL of solid substances
Rotation drying flask
Non-sterile
Glass
B-580 and B-585 ovens
ST/NS neck 24/29
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers