Learn More
BUCHI Valve Unit for Rotavapor RII
Valve unit includes Woulff bottle, 125mL, P+G, holder. Valve is meant to be used with a vacuum controller/interface and non-BUCHI vacuum source
Brand:  BUCHI 047160
Product Code. 10150023
  • £773.00 / Each
Estimated Shipment: 28-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Control house vacuum or 3rd party vacuum pump
  • Connects to BUCHI controllers for vacuum regulation
Specifications
Specifications
Rotavapor RII (use only in combination with a vacuum controller/interface)
Woulff bottle, 125mL; holder
Plastic, glass