Learn More
BINDER™ Buck-Boost Converter
Option for BINDER™ Freezers
Brand:  BINDER™ 8012-1897
Additional Details : Weight : 0.00010kg
Product Code. 16377165
  • £1045.00 / Each
Estimated Shipment: 24-12-2024
to see stock.
Add to basket
Description
Description
  • Buck-boost converter- automatic voltage compensation
Specifications
Specifications
Buck-boost converter - automatic voltage compensation
BINDER Chambers UF V 500 and UF V 700