Learn More
Micronova™ BucketBinders™ Bucket Connectors
Bucket connecting hardware for BucketBinder™ Multi Bucket System
Brand: Micronova™ BB-01
Additional Details : Weight : 1.00000kg
Description
The BucketBinder™ kit includes 2 or 3 polypropylene buckets with casters and one or two sets of BucketBinders (bucket connectors). The system offers easier cleaning and storage - in both small and large controlled environments and the sterile core.Specifications
Bucket Connector, Hardware | |
Autoclavable | |
B-7AC Rectangular Bucket | |
Bucket Connector | |
Non-sterile |
Stainless Steel | |
Cleanroom Surface Cleaning and Disinfecting | |
25 cm | |
1 Pair |