Learn More
Thermo Scientific™ Buckets and Lids for Thermo Scientific Sorvall™ Legend™ Mach 1.6™ 4-Place Swinging Bucket Rotor 75-002-000
For use with Sorvall Legend Mach 1.6/1.6R Centrifuges when equipped with 4-Place Swinging Bucket Rotor 75-002-000.
Brand:  Thermo Scientific™ 75002002
 View more versions of this product
Product Code. 10317552
  • £426.34 / Each
Explore more special offers
Description
Description
For use with Sorvall Legend Mach 1.6/1.6R Centrifuges when equipped with 4-Place Swinging Bucket Rotor 75-002-000.
Choose rectangular or round buckets.
  • Rectangular Bucket (75-002-001) capacity: 150mL
  • Round Bucket (75-002-002) capacity: 400mL
  • Aerosol-tight lids are suitable for biocontainment use
Order Info
Buckets and lids are sold separately. Buckets are sold individually; order correct quantity for your rotor. Lids are sold in pairs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TTH-400 swing bucket rotor
400 mL
400 mL
400mL Round Bucket