missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Budesonide, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Synthetic anti-inflammatory glucocorticoid that displays chemopreventive activity. Prevents formation of lung adenomas and adenocarcinomas in mice following inhalation or oral administration. Reverses DNA hypomethylation and modulates expression of cancer related genes.
Specifications
Specifications
CAS | 51333-22-3 |
Molecular Formula | C25H34O6 |
MDL Number | MFCD00083259,MFCD00138154 |
Quantity | 50 mg |
Synonym | (11β,16α)-16,17-[Butylidenebis(oxy)]-11,21-dihydroxypregna-1,4-diene-3,20-dione |
InChI Key | VOVIALXJUBGFJZ-VXKMTNQYSA-N |
SMILES | CCC[C@@H]1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO |
IUPAC Name | (1S,2S,4R,6R,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one |
Molecular Weight (g/mol) | 430.54 |
PubChem CID | 73759663 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'