missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Capsules pH 9, Reagecon™
Click to view available options
Quantity:
50 Capsules
Specifications
Specifications
Color | Purple |
pH | 9 |
Quantity | 50 Capsules |
Safety and Handling
Product Identifier
- pH BUFFER CAPSULES pH 9.00 ± 0.02 @ 25°C
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : sodium carbonate monohydrate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'