Learn More
Buffer concentrate pH 9.00, Buffer concentrate pH 9.00, Honeywell Fluka™
for 500 ml buffer solution, sodium tetraborate / hydrochloric acid
Brand: Honeywell Fluka 38748-1EA
Code : NEW
Additional Details : Weight : 0.08500kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Buffer concentrate | |
9 | |
1 Pc. |