Learn More
Buffer Concentrated Solution BS pH 4.00 (pHtalate), Standard for pH measurement, Fisher Chemical
Brand: Fisher Chemical J/2821C/05
28.21 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 23
Additional Details : CAS Number : 877-24-7 Weight : 0.10000kg
Specifications
Colorless | |
Liquid | |
282720 |
4 | |
100 mL | |
Ampoule |