Learn More
Buffer Concentrated Solution pH2.00 (Glycine), NIST Standard for pH measurement, Fisher Chemical™
Brand: Fisher Chemical J/2800C/05
Code : 23
Additional Details : CAS Number : 56-40-6 Weight : 0.10000kg
Specifications
Colorless | |
2 | |
Liquid | |
1 Ampule |
Safety and Handling
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.