missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Concentrated Solution pH7.00 (Phosphate), NIST Standard for pH measurement, Fisher Chemical
missing translation for 'orderingAttributeHoverText'
Quantity:
6 Ea.
Packaging:
Ampoule
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 6 Ea. |
MDL Number | 217672 |
Packaging | Ampoule |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'