missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Concentrated Solution pH9.2 (Borate), NIST Standard for pH measurement, Fisher Chemical™
Click to view available options
Quantity:
1 Ampule
6 Ampules
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 6 Ampules |
Packaging | Ampoule |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Signal Word
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'