Learn More
Fisherbrand™ Buffer Saver Blocks
Physically reduce volume in positive and negative compartments of gel chamber thus reducing buffer requirements.
£27.55 - £37.55
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11847633
|
Fisherbrand™
|
Sub-Gel Midi-Plus Horizontal Gel Electrophoresis Unit |
£37.55
Pack of 2 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
11867633
|
Fisherbrand™
|
Sub-Gel Midi Horizontal Gel Electrophoresis Unit |
£32.55
Pack of 2 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
11877633
|
Fisherbrand™
|
Sub-Gel Mini Horizontal Gel Electrophoresis Unit |
£27.55
Pack of 2 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
Description
- Economic low gel and buffer volumes
- Available for multiSUB Horizontal Gel Units
Specifications
Buffer Saver Block | |
2 Each |