missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 4.0 (20°C), With fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST, Honeywell Fluka™
With fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST
£28.42 - £220.53
Chemical Identifiers
CAS | 1336-21-6 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |
IUPAC Name | azanium;hydroxide |
SMILES | N.O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15674020
|
Honeywell-Fluka
33643-100ML |
100 mL | |||||||
15604030
|
Honeywell-Fluka
33643-500ML |
500 mL |
£28.42
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15694020
|
Honeywell-Fluka
33643-1L |
1 L |
£42.19
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15614030
|
Honeywell-Fluka
33643-5L-VP |
5 L |
£86.69
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15624030
|
Honeywell-Fluka
33643-6X1L |
6 x 1 L |
£220.53
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15684020
|
Honeywell-Fluka
33643-10L-VP |
10 L |
£135.14
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1336-21-6 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
CHEBI:18219 | |
N.O |
H5NO | |
MFCD00066650 | |
14923 | |
azanium;hydroxide |
Specifications
1336-21-6 | |
H5NO | |
NONH for all modes of transport | |
N.O | |
35.05 | |
CHEBI:18219 |
4.0 | |
MFCD00066650 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
azanium;hydroxide | |
14923 | |
Buffer solution |
Safety and Handling
P280-P305 + P351 + P338-P337 + P313