missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 4.0 (20°C), Red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide, Honeywell Fluka™
59.68 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
5 L
6 x 500 mL
10 L
6 x 1 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 4.0 |
Quantity | 1 L |
UN Number | NONH for all modes of transport |
IUPAC Name | azanium;hydroxide |
Chemical Name or Material | Buffer solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'