missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 9.0 (20°C), Buffer solution pH 9.0 (20°C), Honeywell Fluka™

Product Code. 15674040
Click to view available options
Quantity:
100 mL
500 mL
1 L
5 L
6 x 100 mL
6 x 500 mL
10 L
6 x 1 L

Borax / hydrochloric acid, traceable to SRM from NIST

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6, 7732-18-5
Synonym Sodium tetraborate buffer solution

Specifications

CAS Max % ≤100.0000%, <1.0000%
CAS Min % ≥0.1000%
Density 1 g/cm3
Quantity 6 x 1 L
Solution Type Buffer
MDL Number MFCD00081185
Packaging Poly Bottle
Chemical Name or Material Buffer solution pH 9.0 (20°C), Buffer solution pH 9.0 (20 °C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.