missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 9.180 (25°C), With fungicide, according to DIN 19266, Honeywell Fluka™

Product Code. 15614020 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 15614020

Brand: Honeywell-Fluka 335951L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

With fungicide, according to DIN 19266

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 1336-21-6
pH 9.180
Quantity 1 L
Molecular Formula H5NO
MDL Number MFCD00066650
UN Number NONH for all modes of transport
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name azanium;hydroxide
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Chemical Name or Material Buffer solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.