missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution Ammonia-Ammonium Chloride, Pure, for Water Hardness Testing, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
Description
Buffer Solution Ammonia-Ammonium Chloride, Pure, for Water Hardness Testing
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 1 L |
Packaging | HDPE plastic bottle |
Vapor Pressure | 330mmHg at 20°C |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'