missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution (Borate), pH 10, Color Coded Blue, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

Brand:  JT Baker 5655-01

83.32 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.



Additional Details : Weight : 0.60000kg Transport : UN number : 1814 Chem class : 8 Pack group : III


 View more versions of this product

Product Code. 15577614

  • £87.70 / 500mL
Estimated Shipment: 23-12-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Buffer Solution (Borate), pH 10
500 mL
1L = 1.00kg
Color Coded Blue
Poly Bottle
BAKER ANALYZED™ Reagent
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362-Take off contaminated clothing.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers