missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution, Cs2CO3 for AAS 0.02mg/L HNO3 matrix, Fisher chemical™
Click to view available options
Quantity:
100 mL
500 mL
Specifications
Concentration | 0.02mg/L |
Quantity | 500 mL |
Solvent or Matrix | HNO3 matrix |
Chemical Name or Material | Buffer solution, Cs2CO3 for AAS |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'