missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution Duracal pH 9.21 (25°C),NIST/PTB reference materials, DKD certified, Honeywell Fluka™
250 mL, certified buffer solution traceable to NIST/PTB reference materials, DKD certified
Brand: Honeywell-Fluka 242146-500ML
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Buffer Solution Duracal Ph 9.21 (25°C) | |
Blue | |
NONH for all modes of transport |
Certified buffer solution traceable to NIST/PTB reference materials, DKD certified | |
500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction