missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution Pack pH 4.01, 7.00, 9.21, Mettler Toledo™
75.43 GBP valid until 2024-07-31
Use promo code "21812" to get your promotional price.
Use promo code "21812" to get your promotional price.
Click to view available options
Quantity:
3 x 10 Sachets (20 mL)
Specifications
pH | 4.01, 7.00, 9.21 |
Quantity | 3 x 10 Sachets (20 mL) |
Packaging | Single-Use Sachets |
Traceability to NIST | Yes |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'