Learn More
Buffer Solution, pH 1.679, Reagecon™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Reagecon's pH Buffer Solution at 25°C is tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable, has guaranteed stability throughout its entire shelf life, even after opening the bottle.
Brand: Reagecon 101679
Description
To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.
- Calibrated to a standard precision NIST value
- Accredited to ISO 17025
Specifications
0°C | |
Colorless | |
Liquid | |
Characteristic | |
Fully miscible in water |
100°C | |
1.679 ±0.01 | |
500 mL | |
Yes |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
167724J1 | Certificate of Analysis | 01/02/2025 | 101679 |
167724F1 | Certificate of Analysis | 30/10/2024 | 101679 |
167724B1 | Certificate of Analysis | 21/06/2024 | 101679 |
167724B1 (1) | Certificate of Analysis | 11/06/2024 | 101679 |
167723H1 | Certificate of Analysis | 23/01/2024 | 101679 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.