missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 1.679, Reagecon™

Reagecon's pH Buffer Solution at 25°C is tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable, has guaranteed stability throughout its entire shelf life, even after opening the bottle.

Brand:  Reagecon 42508

 View more versions of this product

Product Code. 11862422

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Calibrated to a standard precision NIST value
  • Accredited to ISO 17025
Specifications

Specifications

0°C
Colorless
Liquid
Characteristic
Fully miscible in water
100°C
1.679 ±0.01
250 mL
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution, pH 1.679, Reagecon™ > Quantity: 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.