missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 10.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 038710.K2

Code : NEW

Additional Details : CAS Number : 1336-21-6 Weight : 0.01000kg


 View more versions of this product

Product Code. 11318987

  • £35.10 / 1 L
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Buffer solution
1336-21-6
H5NO
VHUUQVKOLVNVRT-UHFFFAOYSA-N
azanium;hydroxide
14923
Specpure™
pH 10.00 (±0.01 at 25°C), No Color, NIST Traceable
1 L
MFCD00066650
N.O
35.05
CHEBI:18219
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Buffer solution, pH 10.00 No Color

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only